Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135032.1 | NP_001128504.1 | 4 | 1481 | ||
NM_032181.2 | NP_115557.1 | 4 | 1441 | |||
GenBank mRNA | AF177337.1 | - | 1514 | |||
AK023453.1 | - | 4 | 1441 | |||
BC016157.1 | - | 4 | 1417 | 2989786 | ||
BC063016.1 | - | 6 | 1547 | 5226973 |
Set Membership: |
G-protein coupled receptor |