Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288834.1 | NP_001275763.1 | 3, 4 | 1231 | ||
NM_032207.3 | NP_115583.1 | 3, 4 | 1231 | |||
XM_006722920.3 | XP_006722983.1 | 3, 4 | 1231 | |||
XM_011528354.2 | XP_011526656.1 | 3, 4 | 1231 | |||
XM_017027350.1 | XP_016882839.1 | 3, 4 | 1231 | |||
GenBank mRNA | AK025395.1 | - | 3, 4 | 1129 | ||
AK225118.1 | - | 3, 4 | 1148 | |||
AK225678.1 | - | 3, 4 | 1141 | |||
AK297781.1 | - | 3, 4 | 372 | |||
AK311324.1 | - | 3, 4 | 619 | |||
BC027869.1 | - | 3, 4 | 1103 | 5205152 |