Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032242.3 | NP_115618.3 | 19 | 3812 | ||
XM_011512908.2 | XP_011511210.1 | 20 | 3914 | |||
XM_011512909.1 | XP_011511211.1 | 20 | 4507 | |||
GenBank mRNA | BQ920360.1 | - | 3 | 539 | ||
X87832.2 | - | 19 | 3747 |