Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003057.2 | NP_003048.1 | 6 | 1085 | ||
NM_153187.1 | NP_694857.1 | 6 | 1085 | |||
XM_005267102.4 | XP_005267159.1 | 6 | 1127 | |||
XM_005267103.1 | XP_005267160.1 | 6 | 1127 | |||
XM_005267104.4 | XP_005267161.1 | 6 | 707 | |||
XM_005267105.4 | XP_005267162.1 | 6 | 659 | |||
XM_006715552.1 | XP_006715615.1 | 6 | 1127 | |||
XM_011536074.2 | XP_011534376.1 | 6 | 950 | |||
GenBank mRNA | AK289887.1 | - | 6 | 1043 | ||
BC126364.1 | - | 6 | 1051 | 8992080 | ||
U77086.1 | - | 6 | 1033 | |||
X98332.1 | - | 6 | 1052 |