Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003059.2 | NP_003050.2 | 5 | 1049 | ||
XM_006714675.3 | XP_006714738.1 | 4 | 456 | |||
XM_011543589.2 | XP_011541891.1 | 3 | 841 | |||
XM_017009776.1 | XP_016865265.1 | 5 | 1097 | |||
GenBank mRNA | AB007448.1 | - | 5 | 1030 | ||
BC028313.1 | - | 5 | 1049 | 5186192 | ||
Y09881.1 | - | 5 | 1042 |