Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032430.1 | NP_115806.1 | 14 | 1934 | ||
XM_005259327.3 | XP_005259384.1 | 14 | 2003 | |||
XM_011527395.2 | XP_011525697.2 | 12 | 1368 | |||
GenBank mRNA | AB058714.1 | - | 13 | 1469 | ||
AF479826.1 | - | 16 | 1832 | |||
AF479827.1 | - | 14 | 1934 | |||
AK124999.1 | - | 6 | 903 | |||
AL834275.1 | - | 14 | 1557 | |||
AY458602.1 | - | 14 | 1706 | |||
AY505124.1 | - | 14 | 1657 | |||
BC016681.1 | - | 6 | 968 | 4341838 |