Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146314.1 | NP_001139786.1 | 4 | 1436 | ||
NM_001254753.1 | NP_001241682.1 | 3 | 1197 | |||
NM_032750.2 | NP_116139.1 | 4 | 1630 | |||
GenBank mRNA | AK075112.1 | - | 4 | 1021 | ||
AK092152.1 | - | 1 | 1821 | |||
AK096073.1 | - | 3 | 1197 | |||
AK296599.1 | - | 4 | 691 | |||
AK298022.1 | - | 5 | 769 | |||
AK298813.1 | - | 3 | 1164 | |||
BC007234.1 | - | 4 | 1014 | 2823284 | ||
BC050650.1 | - | 4 | 828 | |||
BC056411.1 | - | 4 | 837 | 6048106 | ||
BC071931.1 | - | 5 | 1249 | |||
FJ224306.1 | - | 857 |