Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMGNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032806.5 | NP_116195.2 | 2 | 1166 | ||
XM_005265515.2 | XP_005265572.1 | 3 | 1297 | |||
XM_011534163.2 | XP_011532465.1 | 3 | 1285 | |||
XM_011534164.1 | XP_011532466.1 | 2 | 1867 | |||
XM_017007353.1 | XP_016862842.1 | 4 | 1376 | |||
GenBank mRNA | AK027472.1 | - | 1 | 426 | ||
AK092147.1 | - | 2 | 954 | |||
AK124737.1 | - | 1 | 2134 | |||
BC060861.1 | - | 2 | 1156 | 30335722 |