Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033200.2 | NP_149977.2 | 14 | 2397 | ||
XM_005261955.3 | XP_005262012.1 | 14 | 2479 | |||
XM_006724426.3 | XP_006724489.1 | 13 | 2843 | |||
XM_006724427.3 | XP_006724490.1 | 14 | 2394 | |||
XM_017029077.1 | XP_016884566.1 | 13 | 2417 | |||
GenBank mRNA | AK130818.1 | - | 7 | 1208 | ||
BC002942.2 | - | 14 | 2371 | 3954961 | ||
BC014652.1 | - | 14 | 2460 | 4562676 | ||
BC021143.1 | - | 14 | 2371 | 4872642 |