Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272053.1 | NP_001258982.1 | 3 | 1946 | ||
NM_001272054.1 | NP_001258983.1 | 3 | 1993 | |||
NM_033296.2 | NP_150638.1 | 2 | 2065 | |||
GenBank mRNA | AA486335.1 | - | 1 | 219 | ||
AK057736.1 | - | 3 | 1419 | |||
AK074496.1 | - | 2 | 1472 | |||
AK123360.1 | - | 3 | 1946 | |||
AK129651.1 | - | 3 | 1350 | |||
AL539167.3 | - | 1 | 524 | |||
BG255189.1 | - | 1 | 228 | |||
BI858568.1 | - | 1 | 613 |