Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033318.4 | NP_201575.3 | 3 | 773 | ||
GenBank mRNA | AK095636.1 | - | 1 | 787 | ||
BC024237.2 | - | 3 | 769 | 4798793 |