Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199196.1 | NP_001186125.1 | 7 | 1514 | ||
NM_033415.3 | NP_219483.1 | 6 | 1406 | |||
XM_005260157.3 | XP_005260214.1 | 6 | 1457 | |||
XM_017027484.1 | XP_016882973.1 | 6 | 1381 | |||
XM_017027485.1 | XP_016882974.1 | 8 | 1638 | |||
GenBank mRNA | AK295488.1 | - | 6 | 1180 | ||
AK303914.1 | - | 4 | 1094 | |||
BC003700.1 | - | 2 | 152 | |||
BC056161.1 | - | 8 | 1136 | |||
BC067125.1 | - | 7 | 1283 | 6161112 | ||
BX648486.1 | - | 6 | 1301 | |||
CR749678.1 | - | 3 | 747 |