Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052848.1 | NP_443080.1 | 5 | 2620 | ||
XM_005259377.4 | XP_005259434.1 | 5 | 2983 | |||
GenBank mRNA | AK097586.1 | - | 3 | 1783 | ||
AL833726.1 | - | 4 | 2455 | |||
BC007575.1 | - | 3 | 1990 | |||
BC011577.1 | - | 5 | 2620 | 4651484 | ||
BC019597.1 | - | 1 | 467 |