Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135911.1 | NP_001129383.1 | 5 | 1766 | ||
NM_052880.4 | NP_443112.2 | 6 | 1845 | |||
GenBank mRNA | AK074688.1 | - | 6 | 1791 | ||
AK093768.1 | - | 4 | 2354 | |||
AK226089.1 | - | 6 | 1725 | |||
BC011049.2 | - | 6 | 1790 | 4180134 | ||
BC041903.1 | - | 4 | 2142 |