Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHACTR3 SIRNA Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
UniGene ID:
Hs.473218
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125844
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHACTR3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199505.1
NP_001186434.1
11, 12
1586
NM_001199506.1
NP_001186435.1
11, 12
1581
NM_001281507.1
NP_001268436.1
11, 12
1685
NM_080672.4
NP_542403.1
11, 12
2063
NM_183244.1
NP_899067.1
11, 12
1706
NM_183246.1
NP_899069.1
10, 11
1496
XM_011528525.2
XP_011526827.1
11, 12
1692
XM_011528526.2
XP_011526828.1
10, 11
1498
XM_017027626.1
XP_016883115.1
11, 12
1795
XM_017027627.1
XP_016883116.1
11, 12
1740
XM_017027628.1
XP_016883117.1
10, 11
1388
XM_017027629.1
XP_016883118.1
10, 11
1531
XM_017027630.1
XP_016883119.1
10, 11
1359
XM_017027631.1
XP_016883120.1
11, 12
1622
GenBank mRNA
AB098521.1
-
11, 12
1706
AB098522.1
-
10, 11
1496
AJ311122.2
-
11, 12
2062
AJ617581.1
-
11, 12
1581
AK091632.1
-
11, 12
1708
AK098788.1
-
11, 12
1586
AK295459.1
-
11, 12
1685
AK314493.1
-
11, 12
1793
AK316047.1
-
11, 12
1578
BC108303.1
-
11, 12
2063
4179728
BC117362.1
-
11, 12
1951
40125913
BC117364.1
-
11, 12
1954
40125915
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_214459, AY500157.
This siRNA also targets the following transcripts in Mouse: NM_001007154, NM_001177791, NM_028806, NM_001177790, NM_001177789, and others.
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline