Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142650.1 | NP_001136122.1 | 12, 13 | 1636 | ||
NM_138394.3 | NP_612403.2 | 11, 12 | 1892 | |||
XM_005264639.3 | XP_005264696.2 | 11, 12 | 1889 | |||
XM_005264640.3 | XP_005264697.2 | 12, 13 | 1635 | |||
XM_011533165.2 | XP_011531467.1 | 10, 11 | 1817 | |||
XM_011533166.2 | XP_011531468.1 | 11, 12 | 1889 | |||
GenBank mRNA | AK000155.1 | - | 12, 13 | 1540 | ||
AK291462.1 | - | 11, 12 | 1757 | |||
AK292259.1 | - | 12, 13 | 1629 | |||
AL512692.1 | - | 11, 12 | 1369 | |||
AY236962.1 | - | 10, 11 | 1619 | |||
AY236963.2 | - | 12, 13 | 1481 | |||
BC017480.1 | - | 11, 12 | 1738 | 3502860 | ||
KU178839.1 | - | 11, 12 | 1482 | |||
KU178840.1 | - | 11, 12 | 1482 | |||
KU178841.1 | - | 12, 13 | 1565 |