Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011655.2 | NP_001011655.1 | 3 | 503 | ||
NM_001166305.1 | NP_001159777.1 | 3 | 503 | |||
NM_001166306.1 | NP_001159778.1 | 3 | 503 | |||
NM_138399.4 | NP_612408.3 | 3 | 503 | |||
XM_005269371.4 | XP_005269428.1 | 3 | 517 | |||
XM_011513318.2 | XP_011511620.1 | 3 | 406 | |||
XM_011513319.2 | XP_011511621.1 | 3 | 417 | |||
XM_011513320.2 | XP_011511622.1 | 3 | 527 | |||
XM_011513321.2 | XP_011511623.1 | 3 | 414 | |||
XM_011513322.2 | XP_011511624.1 | 3 | 524 | |||
XM_017007517.1 | XP_016863006.1 | 3 | 405 | |||
XM_017007518.1 | XP_016863007.1 | 3 | 400 | |||
GenBank mRNA | AK126914.1 | - | 3 | 242 | ||
AL547748.3 | - | 3 | 514 | |||
BC014883.1 | - | 3 | 363 | 3907191 | ||
BC111485.1 | - | 3 | 462 | 5767916 | ||
BC130297.1 | - | 3 | 407 | 40146328 | ||
BC144158.1 | - | 3 | 407 | 9052678 | ||
BC144159.1 | - | 3 | 407 | 9052679 | ||
BU521517.1 | - | 2 | 59 | |||
BU850042.1 | - | 3 | 466 | |||
BX419619.2 | - | 3 | 464 | |||
BX647418.1 | - | 3 | 262 |