Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135862.2 | NP_001129334.1 | 3, 4 | 235 | ||
NM_001242450.1 | NP_001229379.1 | 2, 3 | 275 | |||
NM_138415.4 | NP_612424.1 | 2, 3 | 280 | |||
NR_134535.1 | 2, 3 | 190 | ||||
XM_006724122.1 | XP_006724185.1 | 2, 3 | 415 | |||
XM_011529855.1 | XP_011528157.1 | 2, 3 | 415 | |||
GenBank mRNA | AK092243.1 | - | 3, 4 | 235 | ||
AK297317.1 | - | 2, 3 | 268 | |||
AK299836.1 | - | 2, 3 | 231 | |||
BC012187.1 | - | 2, 3 | 280 | 4634631 | ||
CR456448.1 | - | 2, 3 | 162 | |||
KU178842.1 | - | 2, 3 | 129 | |||
KU178843.1 | - | 2, 3 | 129 | |||
KU178844.1 | - | 2, 3 | 129 |