Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138797.2 | NP_620152.1 | 8 | 1502 | ||
NR_036556.1 | 8 | 1497 | ||||
XM_006724136.1 | XP_006724199.1 | 7 | 1381 | |||
XM_006724137.1 | XP_006724200.1 | 7 | 1442 | |||
XM_011529877.2 | XP_011528179.1 | 8 | 1925 | |||
XM_011529878.2 | XP_011528180.1 | 8 | 1964 | |||
XM_011529879.2 | XP_011528181.1 | 9 | 1352 | |||
XM_011529880.2 | XP_011528182.1 | 7 | 1357 | |||
XM_011529882.2 | XP_011528184.1 | 8 | 1284 | |||
XM_011529883.2 | XP_011528185.1 | 8 | 1515 | |||
XM_011529884.2 | XP_011528186.1 | 8 | 1317 | |||
XM_017028590.1 | XP_016884079.1 | 8 | 1832 | |||
XM_017028591.1 | XP_016884080.1 | 8 | 1280 | |||
XM_017028593.1 | XP_016884082.1 | 8 | 1767 | |||
GenBank mRNA | AK127583.1 | - | 5 | 1812 | ||
BC014641.1 | - | 8 | 1452 | 2988073 | ||
BC066909.1 | - | 8 | 1469 | 4795922 |