Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321265.1 | NP_001308194.1 | 3, 4 | 392 | ||
NM_001321266.1 | NP_001308195.1 | 2, 3 | 314 | |||
NM_001321267.1 | NP_001308196.1 | 5, 6 | 615 | |||
NM_138799.3 | NP_620154.2 | 4, 5 | 538 | |||
NR_135598.1 | 4, 5 | 538 | ||||
NR_135599.1 | 4, 5 | 538 | ||||
NR_135600.1 | 3, 4 | 392 | ||||
NR_135601.1 | 2, 3 | 314 | ||||
NR_135602.1 | 4, 5 | 538 | ||||
NR_135603.1 | 5, 6 | 615 | ||||
NR_135604.1 | 4, 5 | 538 | ||||
NR_135605.1 | 2, 3 | 314 | ||||
NR_135606.1 | 4, 5 | 538 | ||||
XM_011510313.1 | XP_011508615.1 | 4, 5 | 498 | |||
XM_011510314.2 | XP_011508616.1 | 4, 5 | 498 | |||
GenBank mRNA | AB305044.1 | - | 4, 5 | 402 | ||
AK027321.2 | - | 4, 5 | 498 | |||
AK295907.1 | - | 4, 5 | 446 | |||
AK309818.1 | - | 2, 3 | 156 | |||
AK315881.1 | - | 3, 4 | 321 | |||
BC146871.1 | - | 4, 5 | 482 | 9056799 | ||
BC157827.1 | - | 4, 5 | 482 | 9057046 | ||
BX442603.2 | - | 4, 5 | 538 | |||
DA236556.1 | - | 4, 5 | 451 | |||
DA337762.1 | - | 3, 4 | 307 |