Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321265.1 | NP_001308194.1 | 11 | 1316 | ||
NM_001321266.1 | NP_001308195.1 | 10 | 1238 | |||
NM_001321267.1 | NP_001308196.1 | 13 | 1539 | |||
NM_138799.3 | NP_620154.2 | 12 | 1462 | |||
NR_135598.1 | 13 | 1514 | ||||
NR_135599.1 | 12 | 1781 | ||||
NR_135600.1 | 12 | 1368 | ||||
NR_135601.1 | 10 | 1157 | ||||
NR_135602.1 | 11 | 1329 | ||||
NR_135603.1 | 13 | 1555 | ||||
NR_135604.1 | 12 | 1478 | ||||
NR_135605.1 | 11 | 1290 | ||||
NR_135606.1 | 12 | 1381 | ||||
GenBank mRNA | AB305044.1 | - | 12 | 1326 | ||
AK027321.2 | - | 12 | 1422 | |||
AK074779.1 | - | 2 | 282 | |||
AK309818.1 | - | 10 | 1080 | |||
AK315881.1 | - | 11 | 1245 | |||
BC016005.1 | - | 9 | 1141 | |||
BC146871.1 | - | 12 | 1406 | 9056799 | ||
BC157827.1 | - | 12 | 1406 | 9057046 |