Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM175A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139076.2 | NP_620775.2 | 9 | 1094 | ||
XM_005263280.4 | XP_005263337.1 | 10 | 1227 | |||
XM_017008694.1 | XP_016864183.1 | 8 | 1000 | |||
GenBank mRNA | AK021582.1 | - | 4 | 1302 | ||
AK022704.1 | - | 9 | 1080 | |||
AK023676.1 | - | 8 | 994 | |||
AY358576.1 | - | 9 | 1045 | |||
BC039573.1 | - | 9 | 1041 | 6045433 | ||
EF531340.1 | - | 9 | 1029 |