Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144582.2 | NP_653183.2 | 6 | 3120 | ||
GenBank mRNA | AK056410.1 | - | 2 | 3338 | ||
AK074840.1 | - | 4 | 2770 | |||
AL080223.1 | - | 1 | 1541 | |||
AL832385.1 | - | 1 | 2462 | |||
AY358783.1 | - | 6 | 3080 | |||
BC010656.1 | - | 4 | 2758 | |||
BC020251.1 | - | 4 | 2873 | |||
BG260090.1 | - | 1 | 231 |