Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321343.1 | NP_001308272.1 | 4 | 369 | ||
NM_001321344.1 | NP_001308273.1 | 3 | 242 | |||
NM_001321345.1 | NP_001308274.1 | 2 | 157 | |||
NM_001321346.1 | NP_001308275.1 | 2 | 157 | |||
NM_144632.4 | NP_653233.4 | 2 | 368 | |||
XM_006712287.1 | XP_006712350.1 | 2 | 366 | |||
XM_006712288.3 | XP_006712351.1 | 2 | 405 | |||
XM_011510632.2 | XP_011508934.1 | 2 | 366 | |||
XM_017003375.1 | XP_016858864.1 | 2 | 405 | |||
XM_017003376.1 | XP_016858865.1 | 2 | 153 | |||
GenBank mRNA | AK054856.1 | - | 2 | 128 | ||
AK129862.1 | - | 2 | 327 | |||
AL832688.1 | - | 3 | 239 | |||
AY358262.1 | - | 2 | 466 | |||
BC073844.1 | - | 2 | 104 | |||
BC107694.1 | - | 2 | 112 | |||
BC130306.1 | - | 2 | 315 | 40146337 | ||
BC130308.1 | - | 2 | 312 | 40146339 | ||
CR936731.1 | - | 2 | 344 | |||
CR936732.1 | - | 2 | 340 | |||
CR936735.1 | - | 2 | 367 | |||
CX163622.1 | - | 2 | 97 | |||
DA355461.1 | - | 2 | 155 | |||
DA908857.1 | - | 4 | 367 | |||
HY012093.1 | - | 2 | 157 |