Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003092.4 | NP_003083.1 | 3 | 415 | ||
NM_198220.2 | NP_937863.1 | 3 | 315 | |||
GenBank mRNA | AB451234.1 | - | 2 | 179 | ||
AB451359.1 | - | 2 | 179 | |||
AK313004.1 | - | 3 | 349 | |||
BC008311.1 | - | 3 | 320 | |||
BC018022.1 | - | 3 | 367 | 4286926 | ||
BC036737.2 | - | 3 | 274 | 5533477 | ||
BF243328.1 | - | 393 | ||||
DA088017.1 | - | 3 | 315 | |||
M15841.1 | - | 3 | 304 |