Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UROC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165974.1 | NP_001159446.1 | 21 | 3067 | ||
NM_144639.2 | NP_653240.1 | 20 | 2887 | |||
GenBank mRNA | AK055862.1 | - | 20 | 2865 | ||
BX108463.1 | - | 1 | 41 |