Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144715.3 | NP_653316.3 | 4 | 1288 | ||
XM_005264889.1 | XP_005264946.1 | 4 | 1288 | |||
XM_011533382.1 | XP_011531684.1 | 3 | 1225 | |||
XM_011533383.2 | XP_011531685.2 | 4 | 396 | |||
XM_011533385.2 | XP_011531687.1 | 5 | 664 | |||
XM_017005741.1 | XP_016861230.1 | 6 | 848 | |||
XM_017005742.1 | XP_016861231.1 | 4 | 1288 | |||
XM_017005743.1 | XP_016861232.1 | 5 | 615 | |||
XM_017005744.1 | XP_016861233.1 | 4 | 527 | |||
XM_017005745.1 | XP_016861234.1 | 5 | 590 | |||
GenBank mRNA | AK097644.1 | - | 4 | 1290 | ||
AK122616.1 | - | 6 | 824 | |||
BC028198.1 | - | 4 | 1254 | 5209454 |