Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303105.1 | NP_001290034.1 | 13 | 1680 | ||
NM_001303106.1 | NP_001290035.1 | 13 | 1675 | |||
NM_144978.2 | NP_659415.1 | 13 | 1680 | |||
NR_130121.1 | 13 | 1800 | ||||
XM_006712344.2 | XP_006712407.1 | 13 | 1665 | |||
XM_006712345.3 | XP_006712408.1 | 13 | 1665 | |||
XM_011510757.2 | XP_011509059.1 | 13 | 1662 | |||
XM_011510758.2 | XP_011509060.1 | 12 | 1642 | |||
XM_011510760.2 | XP_011509062.1 | 12 | 1548 | |||
XM_017003492.1 | XP_016858981.1 | 13 | 1675 | |||
XM_017003493.1 | XP_016858982.1 | 11 | 1521 | |||
XM_017003496.1 | XP_016858985.1 | 13 | 1662 | |||
XM_017003497.1 | XP_016858986.1 | 13 | 1675 | |||
XM_017003501.1 | XP_016858990.1 | 13 | 1574 | |||
XM_017003502.1 | XP_016858991.1 | 13 | 1625 | |||
XM_017003503.1 | XP_016858992.1 | 13 | 1564 | |||
XM_017003504.1 | XP_016858993.1 | 12 | 1579 | |||
GenBank mRNA | AK057307.1 | - | 13 | 1667 | ||
AK125199.1 | - | 13 | 1789 | |||
BC038421.1 | - | 13 | 1636 | |||
BC050579.1 | - | 13 | 1673 | 5764613 |