Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144992.4 | NP_659429.4 | 18 | 2770 | ||
XM_005263897.2 | XP_005263954.1 | 18 | 2735 | |||
XM_006712357.2 | XP_006712420.1 | 17 | 2484 | |||
XM_006712359.3 | XP_006712422.1 | 9 | 1164 | |||
XM_006712360.3 | XP_006712423.1 | 6 | 811 | |||
XM_011510770.1 | XP_011509072.1 | 18 | 2735 | |||
XM_011510771.2 | XP_011509073.1 | 17 | 2609 | |||
XM_011510772.1 | XP_011509074.1 | 18 | 2735 | |||
XM_011510774.1 | XP_011509076.1 | 18 | 2735 | |||
XM_017003560.1 | XP_016859049.1 | 12 | 1601 | |||
XM_017003561.1 | XP_016859050.1 | 12 | 1607 | |||
XM_017003562.1 | XP_016859051.1 | 12 | 1657 | |||
XM_017003563.1 | XP_016859052.1 | 12 | 1607 | |||
XM_017003564.1 | XP_016859053.1 | 18 | 2735 | |||
GenBank mRNA | AK057248.1 | - | 15 | 2271 | ||
AK090786.1 | - | 5 | 606 | |||
AK093084.1 | - | 6 | 737 | |||
AK295743.1 | - | 12 | 1665 | |||
AL832635.2 | - | 19 | 2713 | |||
AL832761.1 | - | 18 | 2791 | |||
AL834173.1 | - | 9 | 1169 | |||
AL834385.1 | - | 8 | 1033 | |||
BC026091.2 | - | 18 | 2585 | |||
BC150653.1 | - | 18 | 2710 | 9057024 |