Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144992.4 | NP_659429.4 | 20 | 3009 | ||
XM_005263897.2 | XP_005263954.1 | 20 | 2974 | |||
XM_006712357.2 | XP_006712420.1 | 19 | 2723 | |||
XM_006712359.3 | XP_006712422.1 | 11 | 1403 | |||
XM_006712360.3 | XP_006712423.1 | 8 | 1050 | |||
XM_011510770.1 | XP_011509072.1 | 20 | 2974 | |||
XM_011510771.2 | XP_011509073.1 | 19 | 2848 | |||
XM_011510772.1 | XP_011509074.1 | 20 | 2974 | |||
XM_017003560.1 | XP_016859049.1 | 14 | 1840 | |||
XM_017003561.1 | XP_016859050.1 | 14 | 1846 | |||
XM_017003562.1 | XP_016859051.1 | 14 | 1896 | |||
XM_017003563.1 | XP_016859052.1 | 14 | 1846 | |||
XM_017003564.1 | XP_016859053.1 | 20 | 2974 | |||
GenBank mRNA | AK090786.1 | - | 7 | 845 | ||
AK093084.1 | - | 8 | 976 | |||
AK098841.1 | - | 2 | 248 | |||
AK295743.1 | - | 14 | 1904 | |||
AL832635.2 | - | 21 | 2952 | |||
AL832761.1 | - | 20 | 3030 | |||
AL834173.1 | - | 11 | 1408 | |||
AL834385.1 | - | 10 | 1272 | |||
BC022028.2 | - | 2 | 239 | |||
BC150653.1 | - | 20 | 2949 | 9057024 | ||
EF491813.1 | - | 2 | 114 |