Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145038.4 | NP_659475.2 | 3 | 380 | ||
XM_005264637.3 | XP_005264694.1 | 3 | 372 | |||
XM_005264638.3 | XP_005264695.1 | 1 | 59 | |||
XM_017005271.1 | XP_016860760.1 | 3 | 288 | |||
GenBank mRNA | AK057222.1 | - | 3 | 376 | ||
AK289953.1 | - | 3 | 363 | |||
AK307094.1 | - | 3 | 354 | |||
AL833892.1 | - | 2 | 114 | |||
BC009488.2 | - | 2 | 105 | |||
DB021259.1 | - | 3 | 380 |