Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152266.4 | NP_689479.1 | 3 | 375 | ||
XM_005259393.3 | XP_005259450.1 | 3 | 322 | |||
GenBank mRNA | AK128668.1 | - | 3 | 342 | ||
BC003535.1 | - | 3 | 295 | 3609326 | ||
BC010170.2 | - | 3 | 303 | 3659284 | ||
BC020247.1 | - | 3 | 323 | 4620233 | ||
CD690094.1 | - | 3 | 348 | |||
DA006539.1 | - | 3 | 350 | |||
KU178830.1 | - | 2 | 232 | |||
KU178831.1 | - | 2 | 174 |