Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039693.2 | NP_001034782.1 | 8 | 1953 | ||
NR_004862.1 | 8 | 1934 | ||||
NR_109905.1 | 5 | 1682 | ||||
NR_109906.1 | 7 | 1846 | ||||
NR_109907.1 | 7 | 1798 | ||||
XM_017003310.1 | XP_016858799.1 | 6 | 1581 | |||
GenBank mRNA | AK095272.1 | - | 7 | 1562 | ||
BC025758.1 | - | 7 | 1593 | |||
BX647677.1 | - | 8 | 1936 |