Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257281.1 | NP_001244210.1 | 5, 6 | 641 | ||
NM_001257282.1 | NP_001244211.1 | 5, 6 | 641 | |||
NM_152383.4 | NP_689596.4 | 5, 6 | 641 | |||
NR_046476.1 | 5, 6 | 641 | ||||
NR_046477.1 | 5, 6 | 641 | ||||
GenBank mRNA | AF443854.1 | - | 5, 6 | 540 | ||
AK094293.1 | - | 5, 6 | 642 | |||
AK295363.1 | - | 5, 6 | 511 | |||
AL834174.1 | - | 3, 4 | 180 | |||
BC026166.1 | - | 5, 6 | 545 | 4822064 | ||
BC036113.1 | - | 5, 6 | 540 | 5297794 | ||
BX648325.1 | - | 5, 6 | 533 |