Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLHC1 SIRNA Assays ›
Gene Symbol:
CLHC1
Gene Name:
clathrin heavy chain linker domain containing 1
Gene Aliases:
C2orf63
Chromosome Location:
Chr.2: 55172547 - 55232563 on Build GRCh38
UniGene ID:
Hs.347014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
126082
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLHC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135598.1
NP_001129070.1
8
1047
NM_152385.2
NP_689598.2
10
1498
XM_005264136.2
XP_005264193.1
9
1366
XM_006711937.3
XP_006712000.2
7
805
XM_006711938.3
XP_006712001.1
4
425
XM_006711939.3
XP_006712002.1
3
297
XM_011532519.2
XP_011530821.1
10
4391
XM_011532520.2
XP_011530822.1
10
1669
XM_011532521.2
XP_011530823.1
11
1825
XM_011532522.2
XP_011530824.1
11
1812
XM_011532523.2
XP_011530825.1
10
1901
XM_011532524.1
XP_011530826.1
9
1283
XM_011532525.2
XP_011530827.1
9
1349
XM_011532527.2
XP_011530829.1
10
4993
XM_011532528.1
XP_011530830.1
9
1893
XM_011532529.2
XP_011530831.1
8
1852
XM_011532530.1
XP_011530832.1
8
1613
XM_011532534.1
XP_011530836.1
8
1232
XM_011532535.2
XP_011530837.1
6
742
XM_017003333.1
XP_016858822.1
8
4193
XM_017003334.1
XP_016858823.1
8
1471
XM_017003335.1
XP_016858824.1
8
4561
XM_017003336.1
XP_016858825.1
9
1982
XM_017003337.1
XP_016858826.1
7
1071
XM_017003338.1
XP_016858827.1
7
1049
XM_017003340.1
XP_016858829.1
8
8493
XM_017003341.1
XP_016858830.1
8
1978
XM_017003342.1
XP_016858831.1
7
1479
XM_017003343.1
XP_016858832.1
7
1695
XM_017003344.1
XP_016858833.1
7
1783
XM_017003345.1
XP_016858834.1
6
1653
XM_017003347.1
XP_016858836.1
5
1521
XM_017003349.1
XP_016858838.1
5
3940
XM_017003350.1
XP_016858839.1
5
1451
GenBank mRNA
AK056000.1
-
3
444
AK315683.1
-
10
1456
BC029502.1
-
10
1498
5270720
BC036908.1
-
8
1047
5166064
BM046729.1
-
4
404
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-xzn5f:80/10.239.160.173:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline