Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321547.1 | NP_001308476.1 | 3 | 482 | ||
NM_001321548.1 | NP_001308477.1 | 3 | 482 | |||
NM_001321550.1 | NP_001308479.1 | 3 | 865 | |||
NM_152387.3 | NP_689600.2 | 3 | 865 | |||
GenBank mRNA | AK055884.1 | - | 3 | 457 | ||
AK290867.1 | - | 3 | 456 | |||
BC045189.1 | - | 3 | 836 | 5296647 | ||
BC059366.1 | - | 3 | 813 | |||
BC067755.1 | - | 3 | 456 | 5299303 | ||
CB960352.1 | - | 3 | 483 | |||
DA343572.1 | - | 3 | 482 |