Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002989.3 | NP_002980.1 | 4 | 791 | ||
XM_011518004.2 | XP_011516306.1 | 4 | 811 | |||
GenBank mRNA | AB002409.1 | - | 4 | 748 | ||
AF001979.1 | - | 4 | 690 | |||
AY358887.1 | - | 4 | 695 | |||
BC027918.1 | - | 4 | 697 | 5226781 | ||
U88320.1 | - | 4 | 706 |