Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529914.2 | XP_011528216.1 | 8 | 848 | ||
XM_011529917.2 | XP_011528219.1 | 8 | 707 | |||
XM_011529918.1 | XP_011528220.1 | 8 | 848 | |||
XM_017028621.1 | XP_016884110.1 | 8 | 729 | |||
XM_017028622.1 | XP_016884111.1 | 8 | 648 | |||
XM_017028624.1 | XP_016884113.1 | 7 | 811 | |||
XM_017028625.1 | XP_016884114.1 | 3 | 199 | |||
GenBank mRNA | AK098703.1 | - | 8 | 673 | ||
BC030013.1 | - | 8 | 458 | 4823215 | ||
BI459310.1 | - | 7 | 416 | |||
CR456341.1 | - | 7 | 435 |