Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKAP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304361.1 | NP_001291290.1 | 4 | 911 | ||
NM_152515.4 | NP_689728.3 | 4 | 970 | |||
XM_011510666.2 | XP_011508968.1 | 3 | 843 | |||
GenBank mRNA | AK292530.1 | - | 4 | 921 | ||
AK302875.1 | - | 4 | 852 | |||
AK316241.1 | - | 3 | 835 | |||
AL832036.1 | - | 4 | 921 | |||
BC036217.1 | - | 4 | 902 | 5270339 |