Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095678.1 | - | 1 | 325 | ||
BC101449.1 | - | 1 | 325 | |||
BC101450.1 | - | 1 | 325 | |||
BC101451.1 | - | 1 | 325 | |||
BC113052.1 | - | 1 | 325 |
Gene Symbol: LINC01123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046110.1 | 2 | 1247 | |||
NR_046111.1 | 2 | 1247 | ||||
GenBank mRNA | AK056084.1 | - | 2 | 1164 | ||
BC101448.1 | - | 1 | 325 |