Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2CR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168216.1 | NP_001161688.1 | 9 | 990 | ||
NM_001168217.1 | NP_001161689.1 | 9 | 990 | |||
NM_001168221.1 | NP_001161693.1 | 9 | 990 | |||
NM_152525.5 | NP_689738.3 | 9 | 990 | |||
XM_006712331.3 | XP_006712394.1 | 9 | 990 | |||
XM_006712332.3 | XP_006712395.1 | 9 | 990 | |||
XM_006712333.3 | XP_006712396.1 | 9 | 990 | |||
XM_006712334.3 | XP_006712397.1 | 9 | 990 | |||
XM_006712335.3 | XP_006712398.1 | 9 | 990 | |||
XM_006712336.3 | XP_006712399.1 | 9 | 990 | |||
XM_011510736.2 | XP_011509038.1 | 9 | 990 | |||
GenBank mRNA | AK058080.1 | - | 9 | 962 | ||
AL833429.1 | - | 9 | 987 | |||
BC030659.2 | - | 9 | 983 | 4838801 | ||
BX648732.1 | - | 9 | 983 |