Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033045.3 | NP_001028217.1 | 3 | 478 | ||
NM_001267050.1 | NP_001253979.1 | 3 | 375 | |||
NM_001267051.1 | NP_001253980.1 | 2 | 329 | |||
NM_152529.6 | NP_689742.4 | 2 | 329 | |||
XM_017003485.1 | XP_016858974.1 | 4 | 520 | |||
XM_017003486.1 | XP_016858975.1 | 2 | 381 | |||
XM_017003487.1 | XP_016858976.1 | 3 | 382 | |||
GenBank mRNA | AK091200.1 | - | 2 | 270 | ||
AK307896.1 | - | 2 | 321 | |||
AK315125.1 | - | 2 | 270 | |||
AL834154.1 | - | 3 | 427 | |||
BC028730.1 | - | 3 | 424 | 4813561 | ||
BC035037.1 | - | 3 | 386 | 5260361 | ||
BC036681.1 | - | 2 | 275 | |||
BI548910.1 | - | 3 | 386 | |||
BX537947.1 | - | 2 | 322 | |||
DC334438.1 | - | 2 | 329 |