Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033045.3 | NP_001028217.1 | 3 | 804 | ||
NM_001267050.1 | NP_001253979.1 | 3 | 701 | |||
NM_001267051.1 | NP_001253980.1 | 2 | 655 | |||
NM_152529.6 | NP_689742.4 | 2 | 655 | |||
XM_017003485.1 | XP_016858974.1 | 4 | 846 | |||
XM_017003486.1 | XP_016858975.1 | 2 | 707 | |||
XM_017003487.1 | XP_016858976.1 | 3 | 708 | |||
GenBank mRNA | AK091200.1 | - | 2 | 596 | ||
AK307896.1 | - | 2 | 647 | |||
AK315125.1 | - | 2 | 596 | |||
AL834154.1 | - | 3 | 753 | |||
BC028730.1 | - | 3 | 750 | 4813561 | ||
BC035037.1 | - | 3 | 712 | 5260361 | ||
BC036681.1 | - | 2 | 601 | |||
BX537947.1 | - | 2 | 648 |