Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152678.2 | NP_689891.1 | 14, 15 | 1342 | ||
XM_006713019.3 | XP_006713082.1 | 14, 15 | 1368 | |||
XM_006713020.2 | XP_006713083.1 | 15, 16 | 1574 | |||
XM_017005864.1 | XP_016861353.1 | 15, 16 | 1577 | |||
GenBank mRNA | AK074156.1 | - | 14, 15 | 1329 | ||
AK092288.1 | - | 9, 10 | 744 | |||
BC040291.1 | - | 14, 15 | 1330 | 4822560 | ||
BC054343.1 | - | 5, 6 | 425 |