Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152678.2 | NP_689891.1 | 17 | 1555 | ||
XM_006713019.3 | XP_006713082.1 | 17 | 1581 | |||
XM_006713020.2 | XP_006713083.1 | 18 | 1787 | |||
XM_017005864.1 | XP_016861353.1 | 18 | 1790 | |||
GenBank mRNA | AK074156.1 | - | 17 | 1542 | ||
AK092288.1 | - | 12 | 957 | |||
BC040291.1 | - | 17 | 1543 | 4822560 | ||
BC054343.1 | - | 8 | 638 |