Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206997.1 | NP_001193926.1 | 5, 6 | 569 | ||
NM_152770.2 | NP_689983.2 | 4, 5 | 518 | |||
XM_017007972.1 | XP_016863461.1 | 5, 6 | 670 | |||
XM_017007975.1 | XP_016863464.1 | 3, 4 | 658 | |||
XM_017007978.1 | XP_016863467.1 | 3, 4 | 535 | |||
GenBank mRNA | AY349359.1 | - | 5, 6 | 520 | ||
BC034296.1 | - | 4, 5 | 518 | 5165558 | ||
DB522959.1 | - | 1, 2 | 77 |