Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206997.1 | NP_001193926.1 | 7 | 724 | ||
NM_152770.2 | NP_689983.2 | 6 | 673 | |||
XM_017007972.1 | XP_016863461.1 | 7 | 825 | |||
XM_017007973.1 | XP_016863462.1 | 6 | 698 | |||
XM_017007975.1 | XP_016863464.1 | 5 | 813 | |||
XM_017007978.1 | XP_016863467.1 | 5 | 690 | |||
GenBank mRNA | AY349359.1 | - | 7 | 675 | ||
BC034296.1 | - | 6 | 673 | 5165558 | ||
DB522959.1 | - | 3 | 232 |