Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278298.1 | NP_001265227.1 | 35 | 6810 | ||
NM_153264.6 | NP_694996.5 | 35 | 6810 | |||
NR_022012.2 | 35 | 6810 | ||||
XM_011512621.2 | XP_011510923.1 | 34 | 6621 | |||
GenBank mRNA | AK093199.1 | - | 1 | 31 | ||
AK123718.1 | - | 3 | 621 | |||
AM906082.1 | - | 3 | 958 | |||
AM906083.1 | - | 1 | 145 | |||
EU085556.1 | - | 35 | 6810 |