Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153376.2 | NP_699207.1 | 1 | 1381 | ||
GenBank mRNA | AK075056.1 | - | 1 | 1319 | ||
BC105055.1 | - | 1 | 1377 | 8144058 | ||
BC112274.1 | - | 1 | 1377 | 8327742 |