Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173084.2 | NP_775107.1 | 3 | 893 | ||
GenBank mRNA | AK291284.1 | - | 3 | 815 | ||
AY159379.1 | - | 3 | 893 | |||
BC109259.1 | - | 3 | 880 | 40025031 | ||
BC109260.1 | - | 3 | 880 | 40025036 | ||
BX537987.1 | - | 2 | 914 |